Learn More
Multiroir Controlec™ Trolley
Product Code. 15548943
Click to view available options
Dimensions (L x W x H):
1100 x 520 x 1020 mm (Overall), 504 x 910 mm (Tray)
1100 x 520 x 950 mm (Overall), 504 x 910 mm (Tray)
480 x 800 x 1000 mm (Overall), 435 x 615 mm (Tray)
480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray)
Color:
Black
Light grey
No. of Shelves:
2
3
Unit Size:
Each
Product Code. 15548943
Brand: Multiroir Controlec™ 65007
The Multiroir multi-purpose plastic trolleys are practicle and functional, ideal for general use, transporting and moving multiple items around the lab or workspace.
  • Aluminum frame with handles at each end
  • Available in two or three tray format
  • On casters for easy manouvrability
  • Available in light grey or black
Specifications
Color Black
Dimensions (L x W x H) 480 x 800 x 950 mm (Overall), 435 x 615 mm (Tray)
Load Bearing Capacity (Metric) 180 kg (Overall)
Material Aluminum Frame, Polypropylene Trays
No. of Shelves 2
Shelf Material Polypropylene
Wheel Type Lockable casters