Learn More
Trovirdine, MedChemExpress
Product Code. 16483466
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Product Code. 16483466
Brand: Medchem Express HY15349
MedChemExpress Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.
Trovirdine inhibits HIV-1 RT with an IC50 of 7 nM when employing heteropolymeric primer/template (oligo-DNA/ribosomal RNA)and dGTP as substrate.
Specifications
Chemical Name or Material Trovirdine
CAS 149488-17-5
Color White
Purity Grade Notes Research
Molecular Formula C13H13BrN4S
Quantity 25 mg
Synonym LY300046
Solubility Information DMSO : 100 mg/mL (296.52 mM; Need ultrasonic)
SMILES S=C(NCCC1=NC=CC=C1)NC2=NC=C(Br)C=C2
Molecular Weight (g/mol) 337.24
Formula Weight 337.24
Percent Purity 98.59%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus