Learn More
Invitrogen™ TrueTag™ Donor DNA Kit, YFP
The Invitrogen TrueTag Donor DNA Kit, YFP, provides a rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments.
Brand:  Invitrogen™ A53807
Product Code. 17876501
  • 572.00 EUR / 10 reactions
Description
  • A rapid solution to drive high-efficiency gene tagging (up to 100% edited cells) with transfection-ready donor DNA for genome editing experiments
  • Kit provides everything needed to produce a high-quality DNA donor to tag the gene with a selection marker and YFP
  • Simply provide two locus-specific primers to add homology arms, PCR-amplify, and purify for a transfection-ready donor in hours, not days
  • One-step PCR amplification to generate donor templates – no cloning required
  • Enrich up to 100% YFP-tagged cells
Each kit is designed to enable users to:
  • Tag the carboxy- (C-) or amino- (N-) terminus of the gene of interest
  • Choose from puromycin or blasticidin selection markers – whatever best suits the cell model
  • Easily remove antibiotic resistance markers with Cre/Lox following cell enrichment
Included in the kit:
  • Four linear donor templates: N-terminus/puromycin resistance, N-terminus/blasticidin resistance, C-terminus/puromycin resistance, C-terminus/blasticidin resistance
  • Phusion Flash High-Fidelity PCR Master Mix
  • PCR cleanup columns and buffers
  • Positive control primers to tag human beta-actin (ACTB)
Intuitive software to rapidly design gene-specific reagents
Target-specific gene editing reagents are required for proper knock-in of your fluorescent tag. The Invitrogen TrueDesign Genome Editor web-based software does the design work for you:
  1. Identifies target gene transcript
  2. Designates insertion site
  3. Designs and enables easy ordering of all necessary components
Required reagents available via TrueDesign software (not included in kit):
  • CRISPR guide RNA or TALENs designed adjacent to N- or C-terminus
  • Oligos to PCR-amplify the donor and add appropriate homology arms
  • Sequencing primers for assessment of CRISPR-Cas9 cleavage efficiency
  • Verification primers to validate proper insertion of the TrueTag DNA Donor
Also required and available for purchase separately or as part of TrueDesign workflow:
  • TrueCut Cas9 Protein v2, TrueCut HiFi Cas9 Protein, or GeneART CRISPR mRNA
  • CRISPRMAX Transfection Reagent or MessengerMAX Transfection Reagent
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cellular Imaging
10 reactions
Special Offers
For Research Use Only. Not for use in diagnostic procedures