missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scharlab Tryptone Glucose Extract Agar (TGE)
Tryptone Glucose Extract Agar is for bacteria enumeration in milk, being a complement to Plate Count Agar.
Brand: Scharlab 01-082-500
This item is not returnable.
View return policy
Description
- AOAC/APHA/BAM/COMPF/FDA/ISO/SMD/SMWW
- Plate count medium for milk and dairy products
- According to the Standard Methods for Dairy Products.
Specifications
Solid | |
Agar | |
36°C ± 2.0°C | |
7.0 ±0.2 | |
Insoluble |
Tryptone Glucose Extract Agar | |
Meat Extract 3.0 g/L, Tryptone 5.0 g/L, D(+) Glucose 1.0 g/L, Agar 15.0 g/L | |
44 ±2 hr. | |
500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Scharlab Tryptone Glucose Extract Agar (TGE) > Tryptone Glucose Extract Agar; Form: Solid
Spot an opportunity for improvement?Share a Content Correction