missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Tryptone Soya Broth, Wide Neck
Click to view available options
Quantity:
10 x 100 mL
Unit Size:
Pack of 10
Description
General purpose bottled media complying with EP/USP specification.
Specifications
Specifications
| ATCC® | ARTG |
| Certifications/Compliance | EP/USP |
| Content And Storage | 2°C to 25°C |
| Country of Origin | UK |
| Media Type | Prepared Media |
| Product Type | Microbiology Media |
| For Use With (Application) | Pharmaceutical Bottled Media |
| Packaging Type | Pk. |
| Quantity | 10 x 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction