Learn More
Masterflex™ Tubing adapter NPT (F) PVDF
Description
- Determine which connector type is needed for your application by contacting free technical application assistance
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Always in stock—most materials and configurations available to ship same day
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Meet USP Class VI for pharmaceutical use and FDA, USDA, and 3A standards for food contact
- All fittings are compatible with Tri-Clamp™ connections
- Excellent chemical resistance; low leachable characteristics
Specifications
Specifications
| Description | Tubing Adapter; , 3/4 in. mini to 1/4 in. NPT (F) |
| For Use With | Masterflex L/S and I/P sanitary tubing connection |
| Outlet Port | Sanitary x NPT(F) |
| Thread Size | 1/4 in. |
| Includes | Connection Type: 3/4 in. mini to 1/4 in. NPT(F) |
| Material (Seal) | PVDF |
| Product Type | Vacuum Tubing |