missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Tuff Tip™ Gel Electrolyte Single Junction pH Electrode

This unbreakable epoxy gelled pH electrode with protected Tuff Tip™ shockproof bulb is ideal for reducing maintenance and shortening the response time.

Brand:  Fisherbrand™ 5733534-003I

Product Code. 11765638

  • 110.24 EUR / Each
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Unbreakable epoxy gelled pH electrode with protected Tuff Tip™ shockproof bulb
  • Ceramic annular junction insensitive to pollution
  • The double-junction electrodes are recommended for loaded solutions
  • Reduced maintenance and short response time
  • Supplied with 1 m cable DIN or BNC
 
Specifications

Specifications

Single Junction pH Electrode
1 m
-5°C to +100°C
110 mm
0 to 13
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers