Learn More
Thermo Scientific™ TurboFect in vivo Transfection Reagent
Product Code. 15391516 Shop All Thermo Scientific Products
Click to view available options
Quantity:
120 μL
Unit Size:
120µL
Product Code. 15391516
Brand: Thermo Scientific™ R0541
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Get efficient in vitro delivery of plasmid DNA to cultured cells with low cytotoxicity.
TurboFect Transfection Reagent is recommended for efficient transfection of plasmid DNA. Distinct from lipid-based formulas, TurboFect is a sterile solution of a proprietary cationic polymer in water. The polymer forms positively-charged complexes with DNA that are both compact and stable. These complexes protect DNA from degradation and facilitate efficient plasmid delivery into eukaryotic cells.
Highlights
  • High transfection efficiency of plasmid DNA into a wide variety of cell types
    • Adherent and suspension cells
    • Differentiated and undifferentiated cells
  • Excellent transfection efficiency in the presence or absence of serum 
  • Ready-to-use reagent - no need to reconstitute, dilute, or manipulate
  • Simple protocol
Review our experimental considerations to improve your plasmid transfection results.
Review our experimental considerations to improve your plasmid transfection results.
TRUSTED_SUSTAINABILITY
Specifications
Classification Animal Origin-Free, Chemically Defined
Cell Type Cells in Vivo
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA
Transfection Technique Chemical Transfection
For Use With (Application) Transfection
Product Line TurboFect
Product Type Transfection Reagent
Quantity 120 μL