missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Tygon™ A-60-G

A-60-G Chemical Dispensing Tubing is engineered to outperform neoprene, EPDM, and other general-purpose tubing used in peristaltic pumps.

Brand:  Saint Gobain Life Sciences™ AFL00053

 View more versions of this product

Product Code. 17973471

  • 478.00 EUR / 15m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • High-performance tubing is resistant to acids and alkali
  • Can withstand extreme temperatures from -75°F to +275°F without cracking or deteriorating
  • Heat sealable and can be joined without fittings for easy installation
  • Unlike typical rubber vacuum tubing, resists cracking and aging after years of exposure
  • Available in standard vacuum sizes that will withstand a full vacuum (29.9 mm Hg) at 73°F
  • Very resistant to weathering and abrasion
  • Outstanding flexural fatigue resistance
  • Low gas permeability versus rubber tubing
  • Ozone (300 pphm) and UV light resistant
  • Applications include: soap and disinfectant dispensing, cleaning chemical transfer, caustic chemical dispensing, plating and etching chemicals, glass and window wash systems, vacuum pumps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Not Autoclavable
19 mm
1 in.
49.2 in.
0.126 in.
Chemical Dispensing Tubing
Black
3.5 in. bend radius
13.9 in. Hg (at 73°F)
135°C
0.748 in.
Thermoplastic
25.4 mm
15 m
3.2 mm
120 lbf/in.
61 (Shore A)
7 psi
275°F
Tubing
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.