missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ismatec™ Tygon™ MHSL Extension Tubing
Click to view available options
Tubing Size:
1.02mm I.D.
Unit Size:
10m
Description
- Plasticizer and oil-free.
- Smooth inner-bore.
- Low sorption maintains fluid and tube integrity.
- Does not impart anything into the pumping medium.
- No release of hazardous materials when properly incinerated.
- Applications
- Acids: Excellent
- Alkaline solutions: Excellent
- Solvents: Good/Excellent
- Complies with: FDA.
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 121°C.
- Gas sterilization with ethylene oxide.
- Sterilization with radiation up to 2.5 mrad.
Specifications
Specifications
| Certifications/Compliance | FDA |
| Autoclavable | Autoclavable |
| Material | Tygon MHSL 2001 |
| Overall Length (Metric) | 15 m |
| For Use With (Application) | MEK and other aggressive solvents. |
| Min. Operating Temperature (Metric) | -73°C |
| Tubing Size | 1.02mm I.D. |
| Gas Permeability | CO2 1140, O2 76, N2 190 |
| Color | Transparent |
| Max. Operating Temperature (Metric) | 57°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction