missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Tygon™ ND 100-65 Medical Tubing

The ND 100-65 Tubing is made for blood contact medical devices where non-DEHP plasticized materials are required.

Brand:  Saint Gobain Life Sciences™ ADF00006

Product Code. 17923471

  • 36.90 EUR / 15m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Crystal clear for easy visualization of fluid flow
  • For contact with bodily fluids
  • Non-wetting surface permits complete drainage
  • Non-hemolytic and non-pyrogenic
  • Withstands EtO, gas and gamma sterilization
  • Applications include: minimally invasive devices, blood and IV solutions, dialysis equipment, wound drainage, inhalation equipment, chemotherapy drug delivery, kidney dialysis pumps, peristaltic pumps, catheters
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tubing
0.126 in.
PVC
4.8 mm
15 m
0.7 mm
185 lbf/in.
65 (Shore A)
30 psi
165°F
Autoclavable
3.2 mm
0.189 in.
49.2 in.
0.028 in.
Medical Device Manufacturing
Clear
13 mm bend radius
25 in. Hg (at 73°F)
74°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.