Learn More
Keystone Cleanroom Products Tyvek™ Drawstring Closure Autoclave Bag
Drawstring Closure Autoclave Bag - Designed for the steam sterilization of large or oddly shaped pharmaceutical equipment change parts.
Brand: Keystone Cleanroom Products BGT2A2622D
This item is not returnable.
View return policy
Description
- Designed for steam sterilization
- Tyvek™ on both sides for maximum breathability during sterilization
- Eliminates need to cut material and the use of autoclave tape
- Fast and consistent for equipment preparation area
- Creates secondary level of protection during aseptic presentation of equipment
- Sealed seams sliminate sewing threads and greatly reduce particulates
- Resists tearing and punctures
- Superior microbial barrier
- Lint-free Category 1 (Helmke Drum)
- Manufactured and packaged in a ISO 6 cleanroom environment
- Certificates of Conformance sent with shipment
Specifications
Autoclavable | |
15.4 L | |
Drawstring | |
Single-use | |
Steam Indicator | |
Non-sterile | |
Yes | |
Steam | |
5.30 mil | |
22 in. | |
26 in. |
4.06 gal. | |
Certificate of Conformance | |
White | |
Steam Sterilization of Pharmaceutical Equipment | |
Tyvek | |
0.130 mm | |
250 | |
121°C to 125°C | |
26 x 22 in. | |
55.9 cm | |
66 cm |