Learn More
DuPont™ Tyvek™ IsoClean Disposable Shoe Covers
Product Code. 15590611
Click to view available options
Size:
Medium
Large
IsoClean overshoes are an ideal balance of protection, durability, and comfort.
  • Fixation ties
  • Elastics at ankles and gripper slip-retardant full sole
  • Tyvek™ garments and accessories are composed of flash spun high-density polythylene, providing an ideal balance of protection, durability and comfort
  • Tyvek™ is permeable to both air and water vapor, yet repels water-based liquids and aerosols
  • Garments and accessories made of Tyvek™ IsoClean are used in the biotech, pharmaceutical, medical device manufacturing and electronics industries
  • Suitable for use in cleanroom class ISO 6 and higher
  • Not clean laundered, not sterilized
TRUSTED_SUSTAINABILITY
Specifications
Size Large
Material Tyvek
Color White
Disposable Single-use
Closure Type Elastic Ankle