Learn More
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs
Shop All DuPont Products
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Unit Size:
Pack of 10
Pack of 50
Description
- Silicon non-added
- Stitched internal seams
- 3 pockets and 5 press stud closures
- Composed of flash spun high density polyethylene
- Provides an idea balance of protection, durability and comfort
- Permeable to both air and water vapor, yet repels water-based liquids and aerosols
- Applications include pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Specifications
Specifications
| Product Type | Chemical Resistant Lab Coat |
| Size | Large |
| Gender | Unisex |
| Material | HDPE |
| Closure Type | Press Studs |
| Disposable | Limited-use |
| Sterility | Non-sterile |
| Resistance | Chemicals |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Documents
Scientific Resources
Brochures
Applications and solutions series: Safety edition
File size: 7.7 MB
Flyers
DPT007 Tyvek accessories flyer UK
File size: 515.8 kB
-
Multiple Manufacturers
Product Content Correction
Product Title
DuPont™ Tyvek™ 500 Laboratory Coats Press Studs >
Spot an opportunity for improvement?Share a Content Correction