Learn More
DuPont™ Tyvek 500 Trousers
Composed of flash spun high density polyethylene. DuPont™ Tyvek™ 500 Trousers offer an excellent barrier against fine particles and fibers down to 1 micron in size.
Brand:  DuPont™ D13395741
221.66 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
 View more versions of this product
Product Code. 11743123
  • 252.00 EUR / Pack of 50
Estimated Shipment: 07-05-2025
Description
  • Flash-spun HDPE construction
  • Elasticated waist
  • Stitched seams
  • No pockets
  • White
  • PPE Category I
  • Antistatic treatment (EN 1149-5) - on both sides
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Large
Flashspun HDPE
CE Certified, EN 1149-5
Applications includes pharmaceutical handling, chemical protection, lead and asbestos abatement/remediation, general maintenance/operations, spray painting and general clean-up
Chemical-resistant Trousers
White
Elastic
Internal Stitched Inseam
Special Offers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
    New Offer
    Shop Now
Product Content Correction
Product Title
DuPont™ Tyvek 500 Trousers > Size: Large