Learn More
SI Analytics™ Ubbelohde Viscometers, ASTM
Shop All SI Analytics Products
Click to view available options
Viscosity Range:
0.35 to 1 cSt
200 to 1000 cSt
2000 to 10,000 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Approximate Constant:
Capillary No. 0, Constant K = 0.001
Capillary No. III, Constant K = 1
Capillary No. IIIc, Constant K = 3
Capillary No. IV, Constant K = 10
Capillary No. IVc, Constant K = 30
Diameter (Metric) Opening:
0.24 mm
1.83 mm
2.43 mm
3.27 mm
4.32 mm
Unit Size:
Each
Description
- In accordance with ISO 3105, ASTM D 2515, ASTM D 446
- Calibrated for manual measurements
- Suspended ball level for determination of absolute and relative kinematic viscosity of liquids with Newtonian flow behavior
- Delivered with manufacturer’s certificate in accordance with DIN 55 350, Part 18
- Provided with ring marks for automatic or manual measurements
- Filling quantity: 15 to 20mL
- Overall length: approx. 285mm
- Recommended minimum flowthrough time is 200 seconds
Specifications
Specifications
| Product Type | Ubbelohde ASTM Viscometer |
| Viscosity Range | 0.35 to 1 cSt |
| Calibrated | Yes |
| Approximate Constant | Capillary No. 0, Constant K = 0.001 |
| Certifications/Compliance | ISO 3105, ASTM D 2515, ASTM D 446 |
| Diameter (Metric) Opening | 0.24 mm |
| Length (Metric) | ∼285 mm |
| For Use With (Application) | Determining the viscosity of Newtonian liquids |
| Volume (Metric) | 15 to 20 mL |