Learn More
UDP-GlcNAc Disodium Salt, MedChemExpress
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Description
UDP-GlcNAc Disodium Salt (UDP-α-D-N-Acetylglucosamine Disodium Salt) is a donor substrate of O-GlcNAc transferase (OGT).
Specifications
Specifications
| Chemical Name or Material | UDP-GlcNAc Disodium Salt |
| CAS | 91183-98-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H25N3Na2O17P2 |
| Quantity | 5 mg |
| Synonym | UDP-α-D-N-Acetylglucosamine Disodium Salt |
| Solubility Information | DMSO : 250 mg/mL (383.84 mM; Need ultrasonic) ∣H2O : 125 mg/mL (191.92 mM; Need ultrasonic) |
| SMILES | O[C@H]1[C@H](N2C(NC(C=C2)=O)=O)O[C@H](COP(OP(O[C@@H]3[C@@H]([C@H]([C@H](O)[C@@H](CO)O3)O)NC(C)=O)(O[Na])=O)(O[Na])=O)[C@H]1O |
| Molecular Weight (g/mol) | 651.32 |
| Show More |