Learn More
B Braun™ Sterican™ Single-use Needles
Manufactured from stainless chromium nickel-steel with transparent Luer Lock hub made of polypropylene. B Braun™ Sterican™ Single-use Needles have an extremely smooth surface with light silicone coating to provide minimal pain upon puncture.
Brand:  B Braun™ 9180109
Product Code. 10182552
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • „„Needle made of stainless chromium-nickel steel with extremely smooth surface and light silicone coating „„for minimal pain upon puncture.
  • Transparent Luer Lock plastic hub made of polypropylene
  • „„Colour-coded hub identifies the outer diameter of the needle, conforms to ISO 6009
  • „„Latex-free/PVC-free
Specifications
Specifications
Sterile
EN ISO 7864 and DIN 13097
Green
Withdrawal and flushing
Polypropylene
22 mm
Stainless Chromium Nickel Steel
Luer Lock Hub
0.80 mm
21 G
Yes