missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ UNC 3230
Potent and selective PIP5K1C inhibitor; antinociceptive
279.00 EUR - 1090.00 EUR
Specifications
Purity | >99% |
---|---|
Product Type | UNC 3230 |
Molecular Weight (g/mol) | 344.43 |
Inhibitors | UNC 3230 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732998
|
Tocris Bioscience™
5271/10 |
10 mg |
279.00 EUR
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18786119
|
Tocris Bioscience™
5271/50 |
50 mg |
1090.00 EUR
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent and selective PIP5K1C inhibitor (IC50 = 41 nM). Exhibits selectivity for PIP5K1C over PIP5K1A, the PI 3-kinase family and a panel of other kinases. Reduces PIP2 levels and LPA-induced calcium signaling in dorsal root ganglia (DRG) neurons in vitro. Reduces nociception in mouse models of chronic pain.Specifications
>99% | |
UNC 3230 | |
344.43 | |
UNC 3230 |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title