Learn More
Hughes Unheated Emergency Safety Shower
Floor mounted unheated emergency safety shower, suitable for use in all applications.
566.00 EUR - 1030.00 EUR
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | Price | Quantity & Availability | ||||||
17702358
|
Hughes
HSHS0018GS |
Stainless Steel |
1030.00 EUR
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
17712358
|
Hughes
HSHS0018GV |
Galvanized Steel |
566.00 EUR
Each |
Estimated Shipment: 07-05-2025
Log in to see stock. |
||||||
Description
- Floor mounted unheated emergency safety shower
- ANSI compliant model ideal for use in indoor applications not subject to the possibility of freezing or overheating
- Delivers the ANSI recommended 76 liters per minute of potable water
- Available with either galvanised powder coated mild steel (G) or stainless steel (GS) pipework
Specifications
Floor | |
Safety Shower |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hughes Unheated Emergency Safety Shower