Learn More
Cole-Parmer™ Union Threaded Elbow
Threaded fittings offer more permanent installation
75.85 EUR - 172.00 EUR
Specifications
Product Type Union Threaded Elbow
Description NPT(F) Connection
Autoclavable Not Autoclavable
Material PFA
Sterilization Method Ethylene Oxide
View More Specs
Products 4
Product Code Brand Thread Size Price Quantity & Availability  
Product Code Brand Thread Size Price Quantity & Availability  
11848033
View Documents
Cole-Parmer™
06546-32
1/8 in. NPTF x 1/8 in. NPTF
75.85 EUR
Each
Estimated Shipment: 13-03-2025
to see stock.
Add to basket
11868033
View Documents
Cole-Parmer™
06546-34
3/8 in. NPTF x 3/8 in. NPTF
117.00 EUR
Each
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
11878033
View Documents
Cole-Parmer™
06546-35
1/2 in. NPTF x 1/2 in. NPTF
139.00 EUR
Each
Estimated Shipment: 14-02-2025
to see stock.
Add to basket
11888033
View Documents
Cole-Parmer™
06546-36
3/4 in. NPTF x 3/4 in. NPTF This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Specifications
Union Threaded Elbow
Not Autoclavable
Ethylene Oxide
NPT(F) Connection
PFA
Translucent