Learn More
Kinesis™ Plastic Tubing Cutter
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
14.20 EUR
Specifications
For Use With Upchurch™ standard polymer tubing cutter
View More Specs
Products 1
Product Code Brand Color Material Product Type Price Quantity & Availability  
Product Code Brand Color Material Product Type Price Quantity & Availability  
12986259
View Documents
Kinesis™
02016-86
Silver Steel Replacement Blade
14.20 EUR
Pack of 5
In Stock
Description
Spring-loaded guillotine cutters designed specifically for semi-rigid and rigid polymer tubing
  • Built-in guide holes for both 1/16 and 1/8 in. OD tubing
  • Replacement blades included and sold separately
Flat, 90°, burr-free cut
Offers durability, reliability and ease
  • Able to cut through polymers such as PEEK™, FEP, PFA and Tefzel™
  • Specifications
    Specifications
    Upchurch™ standard polymer tubing cutter