Learn More
UVP PhotoDoc-It™ Imaging Systems
Product Code. 11778356
Click to view available options
Model:
LM-20
LM-26
M20V
PhotoDoc-It 70
Unit Size:
Each
Product Code. 11778356
Brand: UVP 97027408
Capture, print and save color images for basic gel documentation with this compact, economical laboratory system. UVP™ PhotoDoc-It™ Imaging Systems provide a low cost imaging solution without the need for film.
  • The digital color camera has a 10 mega pixel resolution - the user can view images directly on the camera or the (optional) color LCD screen.
  • The lightweight metal hood provides a compact darkroom environment.
  • Images are saved to the camera memory card or instantly printed as 4x6 in. low cost, color prints using the budget photo printer included.
  • An Ethidium™ Bromide filter for fluorescent stained gels (580 to 630nm) is included inside the hood and can be changed for other filter types.
  • A combination lock secures the camera to the darkroom. LCD shown in picture is an optional accessory. Dimensions: 337mm x 241mm x 432mm The PhotoDoc-It system is a very low cost solution for capturing, printing and saving color images for basic gel documentation without the need for film.
Specifications
Voltage 230V
Depth (Metric) 241mm
Filter Type EtBr Red
Pixel Size 16 MP
Height (Metric) 432mm
Wavelength 302-365nm
Width (Metric) 337mm
Model LM-20
Range 302-365nm
Packaging Quantity 1Pack
Printer Included Yes
Camera Type DigiCam 70
For Use With (Application) Gel Imaging, Colorimetric Imaging, Fluorescence
Product Type Imaging System