Learn More
BINDER™ Vacuum Pump Connection Kit
This Vacuum Pump Connection Kit is an accessory for use with vacuum pump VAP 5.
Brand: BINDER™ 8012-2029
359.77 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
This item is not returnable.
View return policy
Description
- Vacuum pump connection kit (ATEX)
- For vacuum pump VAP 5
- Consisting of metal hose, 1.5 meter, clamping ring, inner centering ring and protective covers
Specifications
Vacuum pump connection kit (ATEX) - For vacuum pump VAP 5, consisting of 1.5 m metal hose, clamping ring, internal centering ring and protective covers | |
Vacuum Pump Connection Kit | |
1.5 m metal hose, clamping ring, inner centering ring and protective cover |
BINDER Chambers Series VDL | |
Vacuum Pump VAP 5 | |
ATEX |