Learn More
VACUUBRAND™ VARIO™ Chemistry Pumping Unit, PC 3002 VARIO Select
Provides pinpoint vacuum control by adjusting the motor speed. The automatic evaporation mode is perfect for controlling applications such as rotary evaporation. A variety of condensers are available to suit a range of needs.
Brand:  VACUUBRAND™ 20733574
 View more versions of this product
Product Code. 16201782
  • 9760.00 EUR / Each
Estimated Shipment: 17-03-2025
to see stock.
Explore more special offers
Description
  • VACUU·SELECT Vacuum Controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Automatic boiling point detection and adjustment of the vacuum for short process times
  • Compact and efficient, good ultimate vacuum even with open gas ballast valve
  • Very quiet, low-vibration operation and extra long service intervals
  • Excellent environmental compatibility through low energy consumption and efficient solvent recovery
  • PC 3002 VARIO™ is suitable for work with most common solvents
  • Use manual set-point-control for simple processes, run fully automated distillations, or create unique applications with simple drag-and-drop editing
  • VACUU·SELECT Controller detects solvent boiling and automatically adjusts the pump's motor speed to maintain process control during solvent evaporation
  • Glass inlet separator with a robust protective coating, prevents particles and liquid droplets from entering the pump, preventing damage and extending pump life
  • Touch screen display can be operated even when wearing safety gloves
  • Choose from 14 languages
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7 Mbar (Without Gas Ballast), 12 Mbar (With Gas Ballast)
29 kg
30 to 2400 min-1
419 x 243 x 457 mm
PTFE (Partly Carbon Reinforced), ETFE/ECTFE (Partly Carbon Fiber Reinforced), FFKM
43 dBA
0.53 kW
40°C
2.8 m3/hr.
Diaphragm Pump
2
2
200/230 V
VACUU·SELECT
Ready-to-Use Pumping Unit (Completely Mounted with Manual)
CE-certification; II 3/-G IIC T3 x Internal Atm. Only
Peltronic™ Vapor Condenser works without any cooling media
PC 3002 VARIO Select EK Peltronic
Kaltgerätestecker EN 60320
21.5 kg
10°C
50-60 Hz
IP40
Intelligent Vacuum with VARIO™-control, Rotary Evaporator/Parallel Evaporator, Vacuum Concentrators, Gel Dryer
1.1 bar