Learn More
IKA™ VC 10.300 Check Valve
Check valve designed to connect multiple VC 10 vacuum controllers to a vacuum source.
Brand: IKA™ 0020011105
This item is not returnable.
View return policy
Description
- This check valve is needed to connect multiple VC 10 vacuum controllers in series to a vacuum source
- Every vacuum controller requires this check valve
Specifications
Vacuum | |
0.056 kg |