Learn More
IKA™ VC 10.300 Check Valve
Check valve designed to connect multiple VC 10 vacuum controllers to a vacuum source.
Brand:  IKA™ 0020011105
Product Code. 16298471
Description
Description
  • This check valve is needed to connect multiple VC 10 vacuum controllers in series to a vacuum source
  • Every vacuum controller requires this check valve
Specifications
Specifications
Vacuum
0.056 kg