Learn More
Saint Gobain Life Sciences Versilic™ Folding Skirt Stoppers
Product Code. 11734608
Click to view available options
Diameter (Metric) Inner:
12.1 to 15.6 mm
17.7 to 22.1 mm
20.9 to 26.4 mm
27.6 to 36.3 mm
9.8 to 13.2 mm
Unit Size:
Pack of 10
Pack of 20
Pack of 5
Pack of 50
Product Code. 11734608
Brand: Saint Gobain Life Sciences 46702510
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Silicone biologically neutral Versilic™ Folding Skirt Stoppers.
  • Provides complete tightness on glass, metal or plastic containers regardless of their rims.
  • The turnover is molded to grip the outside of the container neck, providing a double seal in conjunction with the annular serrations on the hollow plug.
  • Can be perforated.
  • Remains watertight after several perforations.
  • Recommended for biological applications.
  • Temperatures between -60°C and +220°C.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Inner 20.9 to 26.4 mm
Quantity 10 Pk.
Temperature Range (Metric) -60°C to +200°C
Material Silicone
Product Type Silicone Stopper