missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences™ Versilon™ 2001 Chemical Resistant Tubing

This clear tubing is highly flexible, chemical resistant, long lasting, and plasticizer- and oil-free.

Brand:  Saint Gobain Life Sciences™ AE300003

Product Code. 17943561

  • 105.00 EUR / 15m

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Highly flexible
  • Chemical resistant
  • Long lasting
  • Plasticizer- and oil-free
  • Helps extend pump life and is ideal for a broad range of demanding applications, including fluid transfer systems, peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, food and beverage dispensing, and chemical transfer
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tubing
0.063 in.
Thermoplastic
4.8 mm
15 m
1.6 mm
140 lbf/in.
68 (Shore A)
45 psi
135°F
Not Autoclavable
1.6 mm
0.189 in.
49.2 in.
0.063 in.
Chemical Resistant Tubing
Clear
Flexible
29.9 in. Hg (at 73°F)
57°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.