Learn More
Brady™ Vinyl Aisle Marking Tape
Product Code. 11741625
Click to view available options
Color:
Black
Blue
Green
Red
White
Yellow
Dimensions (L x W):
32.92 m x 50.80 mm
32.92 m x 76.20 mm
Unit Size:
1 roll
Product Code. 11741625
Brand: Brady™ 058202
Only null left
Add to basket
Add to basket
Vinyl Aisle Marking Tape is a quick, easy way to identify aisles and passageways in your facility.
  • Aisle Marking Tape is a quick, easy way to identify aisles and passageways in your facility.
Specifications
Alternate Part Number AMT-2-GREEN
Description Aisle Marking Tape - Green
Dimensions (L x W) 32.92 m x 50.80 mm
For Use With (Application) Used to mark aisles and passageways
Length (English) 108 ft.
Length (Metric) 32.92 m
Packaging Quantity 1 Roll
Temperature Range (English) -40°F to +176°F
Temperature Range (Metric) -40°C to +80°C
Thickness 0.034 mm
Width (English) 2 in.
Width (Metric) 50.8 mm
Adhesive Type Permanent Rubber-based
Color Green
Material Vinyl
Type Floor Marking Tape