Learn More
Thermo Scientific™ Virtuoso™ Multiple Vial Guide
missing translation for 'orderingAttributeHoverText'
For Use With (Equipment):
Virtuoso Vial Identification System Configured with The Virtuoso Multiple Vial Holder
Förpackningsstorlek
Each
Beskrivelse
Ensure accurate vial identification and sample data integrity by using the Thermo Scientific™ Virtuoso™ Vial Identification System. The system can be configured with the Multiple Vial Holder to hold up to 10 vials and the Multiple Vial Guide to move the vials from the holder to the system for printing one vial at a time in an on-demand fashion.
Tekniske data
Tekniske data
| Product Line | Virtuoso |
| Description | Virtuoso Multiple Vial Guide |
| Device Holds | 10 Vials |
| For Use With (Application) | Vial ID and Labelling |
| For Use With (Equipment) | Virtuoso Vial Identification System Configured with The Virtuoso Multiple Vial Holder |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion