Learn More
Poulten Selfe™ Viscometer Holder
Brand:  Poulten Selfe™ 1824/A2
 View more versions of this product
Product Code. 13101103
  • 231.00 EUR / Each
Estimated Shipment: 05-03-2025
Specifications
Viscometer Holder Suitable
50 mm Bath Opening for Suspended Level Sizes 4a to 5