Learn More
SI Analytics™ ViscoSystem™ AVS 470 Viscometer
Shop All SI Analytics Products
Click to view available options
Product Type:
Viscometer
Unit Size:
Each
Description
- Precise, flexible capillary viscometry system
- Allows suction and pressure measurements completely independently of a PC
- Maximum independence and flexibility
- Allows set up of a measuring station that meets your requirements, even under difficult conditions, eg to monitor production or control quality in the polymer and mineral oil industry
- All types of viscosity calculation are already integrated into the device
- PS2 keyboard to enter additional data
- A serial printer can be used to conveniently document your results
- Optional optical or TC version ViscoPump module can be used to adapt the unit at any time
- Use your existing accessories such as thermostats, stands or flow-through coolers with the AVS 470
Specifications
Specifications
| Product Type | Viscometer |
| Description | Automatic viscometer |