missing translation for 'onlineSavingsMsg'
Learn More
Learn More
von Willebrand Factor Mouse, Clone: VWF/1465, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7450-MSM5-P1
This item is not returnable.
View return policy
Specifications
von Willebrand Factor | |
Cytoplasmic | |
VWF/1465 | |
Unconjugated | |
NP_000543 | |
Mouse | |
250kDa | |
Protein A/G Purified | |
RUO | |
7450 | |
IgG1 |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
VWF | |
Recombinant fragment of human vWF protein (aa1815-1939) | |
HUVEC cells or Tonsil | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |