missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Vortioxetine, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Vortioxetine is a inhibitor of 5-HT1A, 5-HT1B, 5-HT3A, 5-HT7 receptor and SERT, with Ki values of 15 nM, 33 nM, 3.7 nM, 19 nM and 1.6 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | Vortioxetine |
| Physical Form | Solid |
| CAS | 508233-74-7 |
| Color | Yellow |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C18H22N2S |
| For Use With (Application) | Neuroscience-Neuromodulation |
| Synonym | Lu AA 21004 |
| Solubility Information | DMSO : 50 mg/mL (167.53 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction