Learn More
Tocris Bioscience™ VU 0238429
Selective positive allosteric modulator of M5 receptors
Brand:  Tocris Bioscience™ 3634/50
 View more versions of this product
Product Code. 18093852
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
VU 0238429 is a selective positive allosteric modulator of M5 receptors (EC50 values are 1.16, >30 and >30 μM at M5, M1 and M3 receptors respectively). Displays no activity at M2 and M4 receptors.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.97
50 mg
351.28
Store at -20°C
VU 0238429
VU 0238429