Learn More
Tocris Bioscience™ VU 0238429
Selective positive allosteric modulator of M5 receptors
Brand:  Tocris Bioscience™ 3634/50
578.12 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
 View more versions of this product
Product Code. 18093852
  • 636.00 EUR / 50mg
Description
VU 0238429 is a selective positive allosteric modulator of M5 receptors (EC50 values are 1.16, >30 and >30 μM at M5, M1 and M3 receptors respectively). Displays no activity at M2 and M4 receptors.
Specifications
Specifications
0.97
50 mg
351.28
Store at -20°C
VU 0238429
VU 0238429
Special Offers
For Research Use Only