Learn More
Tocris Bioscience™ VU 0546110
Selective SLO3 potassium channel inhibitor
Brand:  Tocris Bioscience™ 7956/10
 View more versions of this product
Product Code. 18015852
  • 142.00 EUR / 10mg
Explore more special offers
Description
Description
VU 0546110 is a selective inhibitor of the sperm-specific potassium channel SLO3 (IC50 = 64 μM). It shows 40-fold selectivity over SLO1. It completely blocks heterologous SLO3 currents and endogenous K+ currents. VU 0546110 inhibits sperm hyperpolarisation and prevents its hyperactivated mobility.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1-[4-[[(4-Methoxyphenyl)thio]methyl]-1-piperidinyl]-2-[4-[(1-methylethyl)sulfonyl]phenyl]ethanone
10 mg
C24H31NO4S2
1421523-00-3
Non-selective/Other Potassium Channel Blockers
0.98