Learn More
Thermo Scientific™ Wash Buffer, pH 8.0
For His-tag protein purification
Brand:  Thermo Scientific™ J61918.AP
Product Code. 15424409
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
8
Ambient temperatures
For His-tag protein purification
500 mL
Liquid
Buffer
Product Suggestions