Learn More
SI Analytics™ Waste Basic Set
SI Analytics Waste Basic Set VZ 8606 with 2.00 mL waste bottle.
Brand: SI Analytics™ 1063833
This item is not returnable.
View return policy
Description
- Tubes VZ 8608 and VZ 8609
- 2.000 mL waste bottle
- Bottle top VZ 6240
- PTFE cuff with NS 29
Specifications
2.000 mL Waste Bottle, Bottle Top VZ 6240 | |
PTFE | |
Tubes VZ 8608 and VZ 8609 | |
2.0 kg |