missing translation for 'onlineSavingsMsg'
Learn More
Water Purified Ph.Eur., Solveco Product Code.: 18042193
Estimated Shipment: 09-04-2025
to see stock.
Add to basket

Water Purified Ph.Eur., Solveco

Product Code. 18042193
25 L
Click to view available options
Quantity:
25 L
This item is not returnable. View return policy

Product Code. 18042193

Brand: Solveco Chemicals 1661

Estimated Shipment: 09-04-2025
to see stock.
Add to basket
Add to basket
This item is not returnable. View return policy

Purified water that is produced in accordance with the standards of the European Pharmacopoeia.

Purified water is produced to meet the stringent quality standards set by the European Pharmacopoeia. This ensures that the water is free from impurities and contaminants, making it suitable for use in pharmaceutical manufacturing, laboratory processes, and other critical applications where high-purity water is essential.

The production process follows rigorous guidelines to guarantee that the water consistently meets the required specifications for purity, safety, and effectiveness, providing a reliable foundation for various scientific and medical uses. Purified water can be utilized for the manufacture of non-sterile pharmaceuticals, such as tablets, creams, and oral/topical solutions, as well as for cleaning equipment and maintaining the integrity of cleanrooms.

Specifications

CAS 7732-18-5
Molecular Formula H2O
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 99.99%
Grade Purified
Packaging HDPE Container
Quantity 25 L
Chemical Name or Material Water
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Water Purified Ph.Eur., Solveco >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.