Learn More
Watson-Marlow Manifold Tubing - Marprene
Double pump segment manifold tubing — use one, transfer to the other for double life.
Brand:  Watson-Marlow 979.0038.000
 View more versions of this product
Product Code. 15323277
  • 352.00 EUR / Pack of 6
Estimated Shipment: 27-02-2025
Description
  • Made from autoclavable marprene
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
7/500 in.
Marprene
0.8 mm
6 Pk.
Orange/Green
0.38 mm
0.03 in.
323S/MC peristaltic pump
0.007 to 0.52 (four roller pump), 0.007 to 0.470 (eight roller pump)mL/min.
Double Segment Tubing