Learn More
Watson-Marlow Manifold Tubing - Marprene
Double pump segment manifold tubing — use one, transfer to the other for double life.
Brand:  Watson-Marlow 979.0152.000
 View more versions of this product
Product Code. 17807953
  • 352.00 EUR / Pack of 6
Estimated Shipment: 12-02-2025
Description
  • Made from autoclavable marprene
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/20 in.
Marprene
0.8 mm
6 Pk.
Yellow/Blue
1.52 mm
0.03 in.
323S/MC peristaltic pump
0.24 to 17 (four roller pump), 0.17 to 12 (eight roller pump)mL/min.
Double Segment Tubing