Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 981.0152.000
 View more versions of this product
Product Code. 12772517
  • 169.00 EUR / Pack of 12
Estimated Shipment: 14-02-2025
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/20 in.
PVC
0.8 mm
1.52mm
Yellow/Blue
1.52 mm
0.03 in.
323S/MC peristaltic pump
0.27 to 19 (four roller pump), 0.2 to 14 (eight roller pump)mL/min.
Pump Tubing