Learn More
Watson-Marlow Manifold Tubing - PVC
Double pump segment manifold tubing — use one, transfer to the other for double life
Brand:  Watson-Marlow 981.0142.000
 View more versions of this product
Product Code. 14855171
  • 169.00 EUR / Pack of 12
Estimated Shipment: 14-02-2025
Description
Double pump segment manifold tubing — use one, transfer to the other for double life
  • For use with 400DM2 and 400DM3 pumpheads
  • Can be used with any multichannel cartridges
  • Adjacent cartridges may contain tubing of a different material or size
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/20 in.
PVC
0.8 mm
1.42mm
Yellow
1.42 mm
0.03 in.
323S/MC peristaltic pump
0.24 to 17 (four roller pump), 0.17 to 12 (eight roller pump)mL/min.
Pump Tubing