missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ WesternDot™ 625 Western Blot Kits

The WesternDot 625 Western Blot kits combine the unique properties of Qdot 625 nanocrystals with the high-affinity streptavidin-biotin interaction to allow simple, highly sensitive detection of proteins on western blots.

429.00 EUR

Specifications

Detection Location In-Blot Detection
Color Red
Detection Method Fluorescence
For Use With (Equipment) iBlot, Photographic Imager
Label or Dye Qdot 625
View More Specs

Products 2
Product Code Brand Description Reactivity Price Quantity & Availability  
Product Code Brand Description Reactivity Price Quantity & Availability  
10062623
View Documents
Invitrogen™
W10132
WesternDot™ 625 Goat Anti-Mouse Western Blot Kit Mouse
429.00 EUR
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
 
10469433
View Documents
Invitrogen™
W10142
WesternDot™ 625 Goat Anti-Rabbit Western Blot Kit Rabbit
429.00 EUR
Each
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
 
Description

Description

The WesternDot 625 Western Blot kits combine the unique properties of Qdot 625 nanocrystals with the high-affinity streptavidin-biotin interaction to allow simple, highly sensitive detection of proteins on western blots. The WesternDot kits contain optimized, ready-to-use or ready-to-dilute reagents for sensitive immunodetection of proteins immobilized on nitrocellulose (NC) or polyvinylidene difluoride (PVDF) membranes.

WesternDot 625 Western Blot kits utilize extremely bright Qdot 625 nanocrystals and include reagents for sensitive imaging of proteins probed with mouse or rabbit primary antibodies.

Features include:
• Detection performance and ease-of-use exceed existing ECL-based methods
• Make use of extremely bright Qdot 625 streptavidin conjugate to allow direct imaging of blots while avoiding the need to spend time and money on optimizing multiple exposures to film in the dark
• Contains reagents sufficient for 20 mini-gel blots (7 x 10 cm/blot)

Specifications

Specifications

In-Blot Detection
Fluorescence
Qdot 625
Qdot Nanocrystals
1 kit
Room Temperature
Red
iBlot, Photographic Imager
625 Goat Anti-Mouse Western Blot Kit
Molecular Probes™, WesternDot™
Protein, Cell Extracts
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 6 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.