Learn More
Cytiva Whatman™ Mixed Cellulose Ester Membrane Filters
Whatman™ Mixed Cellulose Ester Membrane Filters are biologically inert and offers high degree of internal surface area for greater product adsorption.
178.19 EUR - 1060.00 EUR
Product Code | Brand | Membrane Type | Pore Size | Diameter (Metric) | Color | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Membrane Type | Pore Size | Diameter (Metric) | Color | Price | Quantity & Availability | ||
11385374
|
Cytiva
10407314 |
ME25/21 | 0.45 μm | 50 mm | White |
590.00 EUR
Pack of 400 |
In Stock
Log in to see stock. |
||
11395364
|
Cytiva
10406870 |
ME25 | 0.45 μm | 47 mm | White |
178.19 EUR
Pack of 100 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11305374
|
Cytiva
10406871 |
ME25 | 0.45 μm | 4.7 cm | White |
1060.00 EUR
Pack of 1000 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11315374
|
Cytiva
10406872 |
ME25 | 0.45 μm | 50 mm | White |
181.00 EUR
Pack of 100 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11305384
|
Cytiva
10407332 |
ME25 | 0.45 μm | 4.7 cm | Black |
535.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11315384
|
Cytiva
10407334 |
ME25 | 0.45 μm | 50 mm | Black |
590.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11325384
|
Cytiva
10407370 |
ME25 | 0.45 μm | 4.7 cm | Green |
575.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11335384
|
Cytiva
10407372 |
ME25 | 0.45 μm | 50 mm | Green |
590.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11385384
|
Cytiva
10409470 |
ME25 | 0.45 μm | 4.7 cm | Green |
179.00 EUR
Pack of 100 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11395384
|
Cytiva
10409472 |
ME25 | 0.45 μm | 50 mm | Green |
181.00 EUR
Pack of 100 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11982006
|
Cytiva
10408712 |
ME24 | 0.2 μm | 4.7 cm | White |
535.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11355384
|
Cytiva
10408714 |
ME24 | 0.2 μm | 50 mm | White |
535.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
11395374
|
Cytiva
10407324 |
ME25 | 0.45 μm | 50 mm | White |
575.00 EUR
Pack of 400 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
11949217
|
Cytiva
10408915 |
ME27 | 0.8 μm | 50 mm | White |
670.00 EUR
Pack of 400 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
11375024
|
Cytiva
10407342 |
ME29 | 0.8 μm | 47 mm | Black |
815.00 EUR
Pack of 400 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
11305034
|
Cytiva
10407615 |
ME30 | 0.8 μm | 50 mm | Black |
775.00 EUR
Pack of 400 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
11979217
|
Cytiva
10409834 |
ME26 | 0.6 μm | 4.7 cm | Black |
575.00 EUR
Pack of 400 |
Estimated Shipment: 07-11-2024
Log in to see stock. |
||
11375374
|
Cytiva
10407312 |
ME25 | 0.45 μm | 47 mm | White |
520.00 EUR
Pack of 400 |
Estimated Shipment: 12-07-2024
Log in to see stock. |
||
Description
- Sterile options available for critical applications
- Contrast for easier particle detection
- Grids are nontoxic and do not inhibit bacterial growth, ensuring sample integrity
- Black plain and black gridded membranes have mix of cellulose nitrate and cellulose acetate
- Higher dirt loading capacity
- Biologically inert with good thermal stability
- No surfactants to contaminate samples
- Uniform microporous structure of membrane gives high flow rates
Specifications
Autoclavable | |
MCE | |
Sterile |