Learn More
DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached
Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
4 mL
4.0 mL
8 mL
12 mL
16 mL
20 mL
24 mL
25 mL
40 mL
Closure Size:
13-425
15-425
18-400
20-400
24-400
8-425
Material (Liner):
14B Rubber
PTFE/14B Rubber
Unit Size:
Pack of 144
Pack of 288
Pack of 72
Description
Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.
- Fabricated from glass tubing for uniform sidewall and bottom thickness
- Vials and caps are autoclavable
- Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
- Cap and Cap Liners
- Solid top, black phenolic caps
- 14B rubber-lined black phenolic screw cap for aqueous samples
- PTFE/14B rubber-lined black phenolic screw cap for organic samples
Order Info
Replacements caps may be purchased separately.
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements.
Specifications
Specifications
| Material (Liner) | 14B Rubber |
| Color | Clear |
| Diameter (Metric) Outer | 19 mm |
| Material | Borosilicate Glass (Low Extractable) |
| Volume (Metric) | 12 mL |
| For Use With (Application) | Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage |
| Thread Style | Screw |
| Volume (English) | 3.2 dr. |
| Closure Material | Phenolic |
| Autoclavable | Autoclavable |
| Show More |