Learn More
DWK Life Sciences Wheaton™ Biostir™ Single Place Magnetic Stirrer
The Wheaton™ Biostir™ Single Place Magnetic Stirrer is ideal for general mixing and difficult to dissolve solutes. It features programmable functionality, remote control, and alarm.
Brand:  DWK Life Sciences W900702-C
Product Code. 11788869
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Single place
  • Programmable, remote control and alarm capable
  • 150 to 1200 rpm
  • Complies with industry standards
  • Two year warranty
TRUSTED_SUSTAINABILITY
Specifications
Specifications
150 to 1200 rpm
230 VAC
Industry Standards including UL, CSA, CE, WEEE and RoHs
Bio-stir
1
Central EU Cord
Two Year Warranty
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products