Learn More
Schmidlin™ Whisper-0 Nitrogen Generator
This nitrogen generator produces a continuous supply of high purity nitrogen.
Brand:  Schmidlin™ DB-WN2-0-80
Encompass_Preferred
 View more versions of this product
Product Code. 17758929
  • 17802.11 EUR / Each
Explore more special offers
Description
Description
  • Increase efficiency and improve safety and performance
  • On-demand supply 24/7
  • Flow rate at 8 barg (116 psig)
  • Purity: LC/MS grade
  • Proprietary membrane technology
  • No noise, no moving parts and no electricity
  • Ideal for all LC/MS applications
Specifications
Specifications
30 cm
130 cm
7 bar
48 cm
>97% at 80 L/min., >98% at 50 L/min.
80 mL/min.
35°C
18.9 in.
Inlet: 3/8 in., Outlet: 1/4 in.