Learn More
Gilac™ White Square Basin
White Square Basin fit for food contact provides storage with excellent mechanical and chemical resistance in the laboratory. Highly durable, easy to clean, and chemically resistant. Easy to stock since they are nestable.
4.20 EUR - 21.35 EUR
Specifications
Type Storage Tub
Material LDPE
Color White
Shape Square
View More Specs
Products 6
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
13469888
View Documents Promotion Details
Gilac™
G605621
1 L
4.20 EUR
Each
11728511
View Documents Promotion Details
Gilac™
G605821
3 L
7.55 EUR
Each
13479888
View Documents Promotion Details
Gilac™
G605921
4.5 L
9.65 EUR
Each
13489888
View Documents Promotion Details
Gilac™
G606021
7 L
15.50 EUR
Each
13499888
View Documents Promotion Details
Gilac™
G606121
10 L
20.40 EUR
Each
13409898
View Documents Promotion Details
Gilac™
G606221
12 L
21.35 EUR
Each
Description
Description
  • Basin fit for food contact
  • Highly durable and chemical resistant
  • Easy to clean
  • Nesting with no suction effect
  • Moulded rim makes it easy to grasp
Specifications
Specifications
Storage Tub
LDPE
White
Square